Anti-ACTA1 Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29054EA 338 EUR
ABNOPAB29054
Anti-ACTA1 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against purified Rabbit Actin.

Recommended Dilutions: Immunocytochemistry Western Blot: 1:2500-1:5000

Type: Primary
Antigen: ACTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR