Anti-SLC12A1 Rabbit Polyclonal Antibody (ATTO 488)
Proveedor: Abnova
ABNOPAB29038EA 545 EUR
ABNOPAB29038
Anti-SLC12A1 Rabbit Polyclonal Antibody (ATTO 488)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
Recommended Dilutions: Western Blot: 1:1000
Type:
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now