Anti-CNTNAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S65-35]

Proveedor: Abnova
ABNOMAB11903EA 545 EUR
ABNOMAB11903
Anti-CNTNAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S65-35]
Anticuerpos
Mouse monoclonal antibody raised against partial recombinant Cntnap1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: CNTNAP1
Clonality: Monoclonal
Clone: S65-35
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR