Anti-SLC9A3 Rabbit Polyclonal Antibody (Biotin)
Proveedor: Abnova
ABNOPAB29029EA 545 EUR
ABNOPAB29029
Anti-SLC9A3 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3.
Recommended Dilutions: Immunocytochemistry Western Blot: 1:1000
Type:
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now