Anti-SLC9A3 Rabbit Polyclonal Antibody (ATTO 390)

Proveedor: Abnova
ABNOPAB29019EA 545 EUR
ABNOPAB29019
Anti-SLC9A3 Rabbit Polyclonal Antibody (ATTO 390)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3.

Recommended Dilutions: Immunocytochemistry Western Blot: 1:1000

Type:
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: ATTO 390
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR