Anti-RAB5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: Abnova
ABNOPAB29009EA 492 EUR
ABNOPAB29009
Anti-RAB5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type:
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR