Anti-RAB5 Rabbit Polyclonal Antibody (ATTO 488)
Proveedor: Abnova
ABNOPAB29002EA 492 EUR
ABNOPAB29002
Anti-RAB5 Rabbit Polyclonal Antibody (ATTO 488)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.
Recommended Dilutions: Immunofluorescence Western Blot: 1:1000
Type:
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now