Anti-RAB5 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29000EA 356 EUR
ABNOPAB29000
Anti-RAB5 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type: Primary
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR