Anti-SOD2 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Proveedor: Abnova
ABNOPAB28998EA 545 EUR
ABNOPAB28998
Anti-SOD2 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Anticuerpos
Rabbit polyclonal antibody raised against Recombinant of Sod2.
Recommended Dilutions: Western Blot: 0.5 ?g/ml
Type:
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now