Anti-NCAM1 Mouse Monoclonal Antibody [clone: SPM128]

Proveedor: Abnova
ABNOMAB12117EA 471 EUR
ABNOMAB12117
Anti-NCAM1 Mouse Monoclonal Antibody [clone: SPM128]
Anticuerpos
Mouse monoclonal antibody raised against NCAM1.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: SPM128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR