Anti-MYOG Mouse Monoclonal Antibody [clone: SPM144]
Proveedor: Abnova
ABNOMAB12116EA 440 EUR
ABNOMAB12116
Anti-MYOG Mouse Monoclonal Antibody [clone: SPM144]
Anticuerpos
Mouse monoclonal antibody raised against MYOG.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: SPM144
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now