Anti-MUC1 Mouse Monoclonal Antibody [clone: SPM132]
Proveedor: Abnova
ABNOMAB12111EA 440 EUR
ABNOMAB12111
Anti-MUC1 Mouse Monoclonal Antibody [clone: SPM132]
Anticuerpos
Mouse monoclonal antibody raised against MUC1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: SPM132
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now