Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM134]
Proveedor: Abnova
ABNOMAB12106EA 471 EUR
ABNOMAB12106
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM134]
Anticuerpos
Mouse monoclonal antibody raised against EPCAM.
Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: SPM134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now