Anti-CK18 Mouse Monoclonal Antibody [clone: SPM510]
Proveedor: Abnova
ABNOMAB12103EA 440 EUR
ABNOMAB12103
Anti-CK18 Mouse Monoclonal Antibody [clone: SPM510]
Anticuerpos
Mouse monoclonal antibody raised against KRT18.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: SPM510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now