Anti-CK17 Mouse Monoclonal Antibody [clone: SPM560]
Proveedor: Abnova
ABNOMAB12102EA 440 EUR
ABNOMAB12102
Anti-CK17 Mouse Monoclonal Antibody [clone: SPM560]
Anticuerpos
Mouse monoclonal antibody raised against Krt17.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CK17
Clonality: Monoclonal
Clone: SPM560
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now