Anti-CK8 Mouse Monoclonal Antibody [clone: SPM538]
Proveedor: Abnova
ABNOMAB12099EA 440 EUR
ABNOMAB12099
Anti-CK8 Mouse Monoclonal Antibody [clone: SPM538]
Anticuerpos
Mouse monoclonal antibody raised against KRT8.
Recommended Dilutions: Immunohistochemistry: 0.5-1.0 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: SPM538
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now