Anti-IGHG1 Mouse Monoclonal Antibody [clone: SPM556]

Proveedor: Abnova
ABNOMAB12093EA 440 EUR
ABNOMAB12093
Anti-IGHG1 Mouse Monoclonal Antibody [clone: SPM556]
Anticuerpos
Mouse monoclonal antibody raised against native IGHG1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IGHG1
Clonality: Monoclonal
Clone: SPM556
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR