Anti-SOD2 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28982EA 545 EUR
ABNOPAB28982
Anti-SOD2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against Recombinant of Sod2.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR