Anti-KDEL Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Proveedor: Abnova
ABNOPAB28979EA 655 EUR
ABNOPAB28979
Anti-KDEL Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host:
Isotype:
Reactivity:
Order Now