Anti-KDEL Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Proveedor: Abnova
ABNOPAB28976EA 545 EUR
ABNOPAB28976
Anti-KDEL Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Order Now