Anti-KDEL Polyclonal Antibody (ATTO 488)

Proveedor: Abnova
ABNOPAB28966EA 772 EUR
ABNOPAB28966
Anti-KDEL Polyclonal Antibody (ATTO 488)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR