Anti-Biotin Mouse Monoclonal Antibody [clone: SPM375]
Proveedor: Abnova
ABNOMAB12071EA 440 EUR
ABNOMAB12071
Anti-Biotin Mouse Monoclonal Antibody [clone: SPM375]
Anticuerpos
Mouse monoclonal antibody raised against Biotin.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: SPM375
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now