Anti-HSP90AB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Abnova
ABNOPAB28959EA 492 EUR
ABNOPAB28959
Anti-HSP90AB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry Immunoprecipitation: 1:80; Western Blot: 1:1000

Type:
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR