Anti-APP Rabbit Polyclonal Antibody (ATTO 488)

Proveedor: Abnova
ABNOPAB28930EA 643 EUR
ABNOPAB28930
Anti-APP Rabbit Polyclonal Antibody (ATTO 488)
Anticuerpos
Rabbit polyclonal antibody against amyloid oligomers.

Recommended Dilutions: Immunohistochemistry: 1:1000-10000; Immunoprecipitation: 1:1000; Western Blot

Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR