Anti-KCNA6 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S19-36]

Proveedor: Abnova
ABNOMAB11834EA 545 EUR
ABNOMAB11834
Anti-KCNA6 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S19-36]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Kcna6.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: KCNA6
Clonality: Monoclonal
Clone: S19-36
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR