Anti-APP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Proveedor: Abnova
ABNOPAB28922EA 545 EUR
ABNOPAB28922
Anti-APP Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anticuerpos
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.
Recommended Dilutions: Dot Blot: 1:1000
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now