Anti-AQP4 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28892EA 440 EUR
ABNOPAB28892
Anti-AQP4 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Aqp4.

Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000

Type: Primary
Antigen: AQP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR