Anti-AQP3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Proveedor: Abnova
ABNOPAB28890EA 545 EUR
ABNOPAB28890
Anti-AQP3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Aqp3.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP3
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR