Anti-TXNDC5 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2E7/7]
Proveedor: Abnova
ABNOMAB11798EA 545 EUR
ABNOMAB11798
Anti-TXNDC5 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2E7/7]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.
Recommended Dilutions: Western Blot: 1:200
Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2E7/7
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now