Anti-CYSRT1 Mouse Monoclonal Antibody [clone: 4E4]

Proveedor: Abnova
ABNOH00375791-M09EA 394 EUR
ABNOH00375791-M09
Anti-CYSRT1 Mouse Monoclonal Antibody [clone: 4E4]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant C9orf169.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CYSRT1
Clonality: Monoclonal
Clone: 4E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR