Anti-CLECL1 Mouse Monoclonal Antibody [clone: 4A12]
Proveedor: Abnova
ABNOH00160365-M02EA 394 EUR
ABNOH00160365-M02
Anti-CLECL1 Mouse Monoclonal Antibody [clone: 4A12]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant CLECL1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CLECL1
Clonality: Monoclonal
Clone: 4A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now