Anti-RNF32 Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00140545-B01PEA 394 EUR
ABNOH00140545-B01P
Anti-RNF32 Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human RNF32 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: RNF32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now