Anti-AQP3 Rabbit Polyclonal Antibody (Biotin)
Proveedor: Abnova
ABNOPAB28885EA 545 EUR
ABNOPAB28885
Anti-AQP3 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Aqp3.
Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000
Type:
Antigen: AQP3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now