Anti-AQP2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: Abnova
ABNOPAB28865EA 545 EUR
ABNOPAB28865
Anti-AQP2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Aqp2.
Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000
Type:
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now