Anti-AQP1 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))

Proveedor: Abnova
ABNOPAB28853EA 545 EUR
ABNOPAB28853
Anti-AQP1 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR