Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2E7/7]

Proveedor: Abnova
ABNOMAB11782EA 440 EUR
ABNOMAB11782
Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2E7/7]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2E7/7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR