Anti-DNMT1 Mouse Monoclonal Antibody (ATTO 594) [clone: 4G11-C7]
Proveedor: Abnova
ABNOMAB11768EA 545 EUR
ABNOMAB11768
Anti-DNMT1 Mouse Monoclonal Antibody (ATTO 594) [clone: 4G11-C7]
Anticuerpos
Mouse monoclonal antibody raised against partial recombinant DNMT1.
Recommended Dilutions: Western Blot: 2-4 ?g/ml; Immunohistochemistry: 1-2 ?g/ml
Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 4G11-C7
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now