Anti-HSPA1A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 2A4]

Proveedor: Abnova
ABNOMAB11759EA 545 EUR
ABNOMAB11759
Anti-HSPA1A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 2A4]
Anticuerpos
Mouse monoclonal antibody raised against recombinant HSPA1A.

Recommended Dilutions: Western Blot: 1:5000; Immunocytochemistry: 1:100; Immunofluorescence: 1:100; Immunohistochemistry: 1:100

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 2A4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR