Anti-HSPA1A Mouse Monoclonal Antibody (ATTO 594) [clone: 2A4]
Proveedor: Abnova
ABNOMAB11750EA 545 EUR
ABNOMAB11750
Anti-HSPA1A Mouse Monoclonal Antibody (ATTO 594) [clone: 2A4]
Anticuerpos
Mouse monoclonal antibody raised against recombinant HSPA1A.
Recommended Dilutions: Western Blot: 1:5000; Immunocytochemistry: 1:100; Immunofluorescence: 1:100; Immunohistochemistry: 1:100
Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 2A4
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now