Anti-BCL11A Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00053335-B01PEA 394 EUR
ABNOH00053335-B01P
Anti-BCL11A Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human BCL11A protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: BCL11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now