Anti-SLC14A2 Rabbit Polyclonal Antibody (ATTO 594)
Proveedor: Abnova
ABNOPAB28824EA 545 EUR
ABNOPAB28824
Anti-SLC14A2 Rabbit Polyclonal Antibody (ATTO 594)
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
Recommended Dilutions: Immunofluorescence Western Blot: 1:1000
Type:
Antigen: SLC14A2
Clonality: Polyclonal
Clone:
Conjugation: ATTO 594
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now