Anti-SLC14A2 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28820EA 440 EUR
ABNOPAB28820
Anti-SLC14A2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type: Primary
Antigen: SLC14A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR