Anti-SCNN1G Rabbit Polyclonal Antibody (Biotin)
Proveedor: Abnova
ABNOPAB28813EA 545 EUR
ABNOPAB28813
Anti-SCNN1G Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Scnn1g.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now