Anti-GRIN1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S308-48]
Proveedor: Abnova
ABNOMAB11720EA 545 EUR
ABNOMAB11720
Anti-GRIN1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S308-48]
Anticuerpos
Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: GRIN1
Clonality: Monoclonal
Clone: S308-48
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now