Anti-CDKN2A Rabbit Polyclonal Antibody (Biotin)

Proveedor: Abnova
ABNOPAB15357EA 655 EUR
ABNOPAB15357
Anti-CDKN2A Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.

Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 2 ?g/ml

Type:
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR