Anti-IL18 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB19122EA 338 EUR
ABNOPAB19122
Anti-IL18 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against full length recombinant IL18.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR