Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1089CT5.3.1]

Proveedor: Abgent
ABGEAM2251BEA 348 EUR
ABGEAM2251B
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1089CT5.3.1]
Anticuerpos
Type: Primary
Antigen: SIRT1
Clonality: Monoclonal
Clone: 1089CT5.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR