Anti-RPS21 Rabbit Polyclonal Antibody

Proveedor: Abgent
ABGEAP20470BEA 348 EUR
ABGEAP20470B
Anti-RPS21 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RPS21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR