Anti-cRBP7 Rabbit Polyclonal Antibody

Proveedor: Cayman Chemical
CAYM10010250-1EA 450 EUR
CAYM10010250-1
Anti-cRBP7 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: cRBP7
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: African Green Monkey, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR