Anti-5-LO Rabbit Polyclonal Antibody

Proveedor: Cayman Chemical
CAYM10007820-1EA 614 EUR
CAYM10007820-1
Anti-5-LO Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: 5-LO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, non-Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR